Assay Details
Gene Symbol: Tex264 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001007665.1 | NP_001007666.1 | 4 - 5 | 733 | 90 | |
XM_006243726.2 | XP_006243788.1 | 4 - 5 | 699 | 90 | ||
XM_006243727.2 | XP_006243789.1 | 4 - 5 | 1000 | 90 | ||
XM_008766501.1 | XP_008764723.1 | 4 - 5 | 689 | 90 | ||
GenBank mRNA | BC083732.1 | - | 4 - 5 | 733 | 90 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
MicroArray Probes:
