Assay Details
Gene Symbol: Parp3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001008328.1 | NP_001008329.1 | 4 - 5 | 610 | 66 | |
GenBank mRNA | BC085863.1 | - | 4 - 5 | 610 | 66 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
