Assay Details
Gene Symbol: Parp3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001008328.1 | NP_001008329.1 | 7 - 8 | 1112 | 81 | |
XM_006243725.2 | XP_006243787.1 | 7 - 8 | 1448 | 81 | ||
XM_008766500.1 | XP_008764722.1 | 7 - 8 | 1479 | 81 | ||
GenBank mRNA | BC085863.1 | - | 7 - 8 | 1112 | 81 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
