Assay Details
Gene Symbol: Parp3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001008328.1 | NP_001008329.1 | 8 - 9 | 1193 | 63 | |
XM_006243725.2 | XP_006243787.1 | 8 - 9 | 1529 | 63 | ||
XM_008766500.1 | XP_008764722.1 | 8 - 9 | 1560 | 63 | ||
GenBank mRNA | BC085863.1 | - | 8 - 9 | 1193 | 63 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
