Assay Details
Gene Symbol: Ppm1m | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_008766595.1 | XP_008764817.1 | 8 - 9 | 1901 | 63 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
