Assay Details
Gene Symbol: Alas1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_024484.2 | NP_077810.2 | 3 - 4 | 530 | 97 | |
GenBank mRNA | BC061793.1 | - | 3 - 4 | 530 | 97 | |
J04044.1 | - | 2 - 3 | 461 | 97 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
