Assay Details
Gene Symbol: Alas1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_024484.2 | NP_077810.2 | 6 - 7 | 1084 | 75 | |
GenBank mRNA | BC061793.1 | - | 6 - 7 | 1084 | 75 | |
J03190.1 | - | 5 - 6 | 1008 | 75 | ||
J04044.1 | - | 5 - 6 | 1015 | 75 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
