Assay Details
Gene Symbol: Alas1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_024484.2 | NP_077810.2 | 8 - 9 | 1427 | 83 | |
GenBank mRNA | BC061793.1 | - | 8 - 9 | 1427 | 83 | |
J03190.1 | - | 7 - 8 | 1351 | 83 | ||
J04044.1 | - | 7 - 8 | 1358 | 83 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
