Assay Details
Gene Symbol: Bfsp2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001277434.1 | NP_001264363.1 | 3 - 4 | 791 | 68 | |
GenBank mRNA | FQ228265.1 | - | 3 - 4 | 415 | 68 | |
FQ230804.1 | - | 1 - 2 | 75 | 68 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() |
MicroArray Probes:
