Assay Details
Gene Symbol: Gnai2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_031035.3 | NP_112297.1 | 8 - 9 | 1248 | 87 | |
GenBank mRNA | BC078806.1 | - | 8 - 9 | 1200 | 87 | |
DQ120471.1 | - | 8 - 9 | 1157 | 87 | ||
DQ120472.1 | - | 8 - 9 | 1157 | 87 | ||
M12672.1 | - | 8 - 9 | 1144 | 87 | ||
M17528.1 | - | 8 - 9 | 1246 | 87 |

More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
