Assay Details
Gene Symbol: Pdgfd | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_023962.2 | NP_076452.1 | 1 - 2 | 234 | 73 | |
GenBank mRNA | AB052170.2 | - | 1 - 2 | 234 | 73 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
