Assay Details
Gene Symbol: Pdgfd | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_023962.2 | NP_076452.1 | 4 - 5 | 685 | 85 | |
GenBank mRNA | AB052170.2 | - | 4 - 5 | 685 | 85 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
