Assay Details
Gene Symbol: Birc3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_023987.3 | NP_076477.3 | 7 - 8 | 1545 | 99 | |
XM_006242499.2 | XP_006242561.1 | 6 - 7 | 3508 | 99 | ||
XM_006242500.2 | XP_006242562.1 | 7 - 8 | 2278 | 99 | ||
XM_006242502.2 | XP_006242564.1 | 6 - 7 | 1777 | 99 | ||
XM_006242503.2 | XP_006242565.1 | 6 - 7 | 1736 | 99 | ||
GenBank mRNA | AF183430.1 | - | 5 - 6 | 1313 | 99 | |
BC083555.1 | - | 7 - 8 | 1545 | 99 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
