Assay Details
Gene Symbol: Yap1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001034002.2 | NP_001029174.2 | 1 - 2 | 274 | 102 | |
XM_006242488.2 | XP_006242550.1 | 1 - 2 | 921 | 102 | ||
XM_006242489.2 | XP_006242551.1 | 1 - 2 | 919 | 102 | ||
XM_006242490.2 | XP_006242552.1 | 1 - 2 | 920 | 102 | ||
XM_006242491.2 | XP_006242553.1 | 1 - 2 | 918 | 102 | ||
XM_006242492.2 | XP_006242554.1 | 1 - 2 | 920 | 102 | ||
XM_006242493.2 | XP_006242555.1 | 1 - 2 | 918 | 102 | ||
XM_008765893.1 | XP_008764115.1 | 1 - 2 | 921 | 102 | ||
XM_008765894.1 | XP_008764116.1 | 1 - 2 | 919 | 102 | ||
GenBank mRNA | DQ186896.2 | - | 1 - 2 | 274 | 102 | |
DQ186897.2 | - | 1 - 2 | 274 | 102 | ||
DQ186898.2 | - | 1 - 2 | 274 | 102 | ||
DQ376007.1 | - | 1 - 2 | 274 | 102 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
