Assay Details
Gene Symbol: Yap1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001034002.2 | NP_001029174.2 | 5 - 6 | 931 | 65 | |
XM_006242490.2 | XP_006242552.1 | 5 - 6 | 1583 | 65 | ||
XM_006242491.2 | XP_006242553.1 | 5 - 6 | 1575 | 65 | ||
GenBank mRNA | DQ186898.2 | - | 5 - 6 | 931 | 65 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
