Assay Details
Gene Symbol: Yap1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_006242492.2 | XP_006242554.1 | 5 - 6 | 1583 | 68 | |
XM_006242493.2 | XP_006242555.1 | 5 - 6 | 1575 | 68 | ||
GenBank mRNA | DQ376007.1 | - | 5 - 6 | 931 | 68 |

More Information
Set Membership: |
![]() ![]() ![]() |
Gene Ontology Categories:
