Assay Details
Gene Symbol: Elavl3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_172324.1 | NP_758827.1 | 1 - 2 | 50 | 114 | |
XM_006242616.2 | XP_006242678.1 | 1 - 2 | 394 | 114 | ||
GenBank mRNA | AB097198.1 | - | 1 - 2 | 50 | 114 |

More Information
Additional Information:
This assay is in the 5'UTR region of the AB097198.1, NM_172324.1, XM_006242616.2 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
