Assay Details
Gene Symbol: Pgr | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_022847.1 | NP_074038.1 | 4 - 5 | 1875 | 88 | |
GenBank mRNA | HM037361.1 | - | 2427 | 88 | ||
HM037362.1 | - | 2427 | 88 | |||
L16922.1 | - | 4 - 5 | 1875 | 88 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
