Assay Details
Gene Symbol: Rasgef1a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_006237211.2 | XP_006237273.1 | 2 - 3 | 309 | 100 | |
XM_008763269.1 | XP_008761491.1 | 2 - 3 | 445 | 100 | ||
XM_232315.7 | XP_232315.6 | 2 - 3 | 471 | 100 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
