Assay Details
Gene Symbol: Nr4a3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_008763702.1 | XP_008761924.1 | 9 | 7304 | 68 | |
XM_008763703.1 | XP_008761925.1 | 8 | 7232 | 68 | ||
GenBank mRNA | DQ268830.1 | - | 8 | 5517 | 68 |

More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
