Assay Details
Gene Symbol: Grin3a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | AF061945.1 | - | 2 - 3 | 253 | 59 | |
AF073379.1 | - | 3240 | 59 | |||
L34938.1 | - | 2767 | 59 | |||
U29873.1 | - | 3240 | 59 |

More Information
Set Membership: |
![]() ![]() ![]() |
