Assay Details
Gene Symbol: Grin3a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | L34938.1 | - | 3008 | 97 | ||
U29873.1 | - | 3481 | 97 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
MicroArray Probes:
Agilent
Affymetrix
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Agilent Whole Rat Genome Microarray 4x44K v2 | A_64_P154352 | - | - |
Agilent Whole Rat Genome Microarray 4x44K v3 | A_64_P154353 | - | - |
Agilent Whole Rat Genome Microarray 4x44K v3 | A_64_P154352 | - | - |
Agilent Whole Rat Genome Microarray 4x44K v3 | A_64_P154352 | - | - |
Agilent Whole Rat Genome Microarray 4x44K v3 | A_64_P154353 | - | - |
Agilent SurePrint G3 Rat GE 8x60K Microarray | A_64_P154352 | - | - |
Agilent SurePrint G3 Rat GE 8x60K Microarray | A_64_P154353 | - | - |
SurePrint G3 Rat Gene Expression Microarray G4131A | A_44_P132873 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 39518 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 64205 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 74563 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 78949 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 97009 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 53308 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 12367 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 26433 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 40759 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 62057 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 99853 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Rat Gene 1.0 ST Array | 10876737 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10876740 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10876742 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10876741 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10876743 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10876745 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10876736 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10876746 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10876735 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10876739 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10876738 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10876744 | - | - |
Affymetrix Rat Gene 1.1 ST Array | 10876734 | - | - |
Rat Genome 230 2.0 Array | 1370601_PM_a_at | - | - |
Rat Genome 230 2.0 Array | 1370601_a_at | - | - |
Affymetrix HT Rat Focus Array | 1370601_a_at | - | - |
Affymetrix HT Rat Focus Array | 1370601_PM_a_at | - | - |
Rat Expression Set 230 | 1370601_a_at | - | - |
Rat Expression Set 230 | 1370601_PM_a_at | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10876734 | - | - |
Rat Genome U34 Set | AF061945_g_at | - | - |
Rat Genome U34 Set | AF061945_at | - | - |
Rat Genome U34 Set | U29873_at | - | - |
Rat Genome U34 Set | AF061945_at | - | - |
Rat Genome U34 Set | U29873_at | - | - |
Rat Genome U34 Set | AF061945_g_at | - | - |
