Assay Details
Gene Symbol: Nr4a3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_031628.1 | NP_113816.1 | 6 - 7 | 2161 | 74 | |
XM_008763702.1 | XP_008761924.1 | 7 - 8 | 3955 | 74 | ||
XM_008763703.1 | XP_008761925.1 | 6 - 7 | 3883 | 74 | ||
GenBank mRNA | D38530.1 | - | 6 - 7 | 2161 | 74 | |
DQ268830.1 | - | 6 - 7 | 2161 | 74 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
