Assay Details
Gene Symbol: Tgfbr1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_012775.2 | NP_036907.2 | 1182 | 146 | ||
XM_006238030.2 | XP_006238092.2 | 1196 | 146 | |||
GenBank mRNA | BC087035.1 | - | 1124 | 146 | ||
L26110.1 | - | 1151 | 146 |
Gene Symbol: LOC103690035 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_006238153.2 | XP_006238215.2 | 5 - 6 | 2467 | 146 | |
XM_008763706.1 | XP_008761928.1 | 5 - 6 | 2479 | 146 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
