Assay Details
Gene Symbol: Sfxn1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001012213.1 | NP_001012213.1 | 9 - 10 | 889 | 93 | |
XM_006253645.2 | XP_006253707.1 | 9 - 10 | 1100 | 93 | ||
XM_008771502.1 | XP_008769724.1 | 9 - 10 | 861 | 93 | ||
GenBank mRNA | BC078935.1 | - | 9 - 10 | 889 | 93 | |
FQ209748.1 | - | 843 | 93 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
