Assay Details
Gene Symbol: Il9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001105747.1 | NP_001099217.1 | 4 - 5 | 338 | 105 | |
GenBank mRNA | L36460.1 | - | 4 - 5 | 352 | 105 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
