Assay Details
Gene Symbol: F12 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001014006.1 | NP_001014028.1 | 13 - 14 | 1858 | 101 | |
GenBank mRNA | BC088187.1 | - | 13 - 14 | 1858 | 101 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
