Assay Details
Gene Symbol: Smad5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_021692.1 | NP_067724.1 | 5 - 6 | 1324 | 80 | |
XM_006253586.2 | XP_006253648.1 | 8 - 9 | 1789 | 80 | ||
XM_006253587.2 | XP_006253649.1 | 7 - 8 | 1707 | 80 | ||
XM_006253588.2 | XP_006253650.1 | 8 - 9 | 2031 | 80 | ||
XM_008771446.1 | XP_008769668.1 | 7 - 8 | 3367 | 80 | ||
XM_008771447.1 | XP_008769669.1 | 8 - 9 | 2549 | 80 | ||
XM_008771448.1 | XP_008769670.1 | 8 - 9 | 2724 | 80 | ||
GenBank mRNA | AB010955.1 | - | 5 - 6 | 1324 | 80 | |
BC161849.1 | - | 6 - 7 | 1511 | 80 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
