Assay Details
Gene Symbol: Lyn | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001111098.1 | NP_001104568.1 | 3 - 4 | 347 | 105 | |
GenBank mRNA | AF000301.1 | - | 3 - 4 | 347 | 105 | |
AF000302.1 | - | 3 - 4 | 345 | 105 | ||
L14823.1 | - | 2 - 3 | 141 | 105 |

More Information
Set Membership: |
![]() ![]() ![]() |
Gene Ontology Categories:
