Assay Details
Gene Symbol: Chchd7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003749892.3 | XP_003749940.1 | 2 - 3 | 410 | 68 | |
XM_003749893.2 | XP_003749941.1 | 2 - 3 | 249 | 68 | ||
XM_008763563.1 | XP_008761785.1 | 2 - 3 | 428 | 68 |

More Information
Additional Information:
This assay is in the 5'UTR region of the XM_003749892.3, XM_003749893.2, XM_008763563.1 transcripts.
Set Membership: |
![]() ![]() ![]() |
