Assay Details
Gene Symbol: Prr14l | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_006251416.2 | XP_006251478.1 | 9 - 10 | 5859 | 131 | |
XM_008770385.1 | XP_008768607.1 | 9 - 10 | 5863 | 131 | ||
XM_008770386.1 | XP_008768608.1 | 8 - 9 | 6501 | 131 | ||
XM_008770387.1 | XP_008768609.1 | 9 - 10 | 6189 | 131 | ||
XM_008770388.1 | XP_008768610.1 | 8 - 9 | 6052 | 131 | ||
XM_223586.7 | XP_223586.5 | 8 - 9 | 5700 | 131 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
MicroArray Probes:
