Assay Details
Gene Symbol: Trmt44 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001130061.1 | NP_001123533.1 | 6 - 7 | 1051 | 103 | |
GenBank mRNA | FQ212309.1 | - | 6 - 7 | 1031 | 103 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
