Assay Details
Gene Symbol: Mxd4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001108364.1 | NP_001101834.1 | 4 - 5 | 564 | 86 | |
XM_006251328.1 | XP_006251390.1 | 5 - 6 | 863 | 86 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
