Assay Details
Gene Symbol: Mxd4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_006251328.1 | XP_006251390.1 | 2 - 3 | 559 | 120 |

More Information
Set Membership: |
![]() ![]() ![]() |
Gene Ontology Categories:
