Assay Details
Gene Symbol: Pacrgl | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_341225.7 | XP_341226.5 | 1 - 2 | 144 | 98 |

More Information
Additional Information:
This assay is in the 5'UTR region of the XM_341225.7 transcripts.
Set Membership: |
![]() ![]() ![]() |
