Assay Details
Gene Symbol: Pacrgl | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_006251057.2 | XP_006251119.1 | 5 - 6 | 611 | 108 | |
XM_008770221.1 | XP_008768443.1 | 5 - 6 | 1419 | 108 | ||
XM_008770222.1 | XP_008768444.1 | 5 - 6 | 1040 | 108 | ||
XM_341225.7 | XP_341226.5 | 5 - 6 | 520 | 108 |

More Information
Set Membership: |
![]() ![]() ![]() |
