Assay Details
Gene Symbol: Cpz | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_031766.1 | NP_113954.1 | 3 - 4 | 566 | 130 | |
XM_006251151.2 | XP_006251213.1 | 2 - 3 | 561 | 130 | ||
GenBank mRNA | AF017637.1 | - | 3 - 4 | 566 | 130 | |
AF017638.1 | - | 3 - 4 | 566 | 130 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
