Assay Details
Gene Symbol: Nr4a2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_006234201.2 | XP_006234263.1 | 1 - 2 | 207 | 114 | |
GenBank mRNA | FQ222109.1 | - | 1 - 2 | 228 | 114 | |
U72345.1 | - | 1 - 2 | 213 | 114 |

More Information
Additional Information:
This assay is in the 5'UTR region of the U72345.1, XM_006234201.2 transcripts.
Set Membership: |
![]() ![]() ![]() |
Gene Ontology Categories:
