Assay Details
Gene Symbol: Nr4a2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_019328.3 | NP_062201.2 | 2 - 3 | 974 | 120 | |
XM_006234201.2 | XP_006234263.1 | 3 - 4 | 1192 | 120 | ||
XM_008761861.1 | XP_008760083.1 | 2 - 3 | 1108 | 120 | ||
GenBank mRNA | DQ092629.1 | - | 2 - 3 | 905 | 120 | |
DQ092630.1 | - | 2 - 3 | 741 | 120 | ||
DQ092631.1 | - | 2 - 3 | 919 | 120 | ||
DQ092632.1 | - | 2 - 3 | 909 | 120 | ||
DQ092633.1 | - | 2 - 3 | 741 | 120 | ||
DQ092634.1 | - | 2 - 3 | 741 | 120 | ||
FQ222109.1 | - | 3 - 4 | 1213 | 120 | ||
L08595.1 | - | 2 - 3 | 974 | 120 | ||
U01146.1 | - | 2 - 3 | 905 | 120 | ||
U72345.1 | - | 3 - 4 | 1198 | 120 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
