Assay Details
Gene Symbol: Lypd6b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001134580.1 | NP_001128052.1 | 8 - 9 | 735 | 68 | |
XM_008761762.1 | XP_008759984.1 | 5 - 6 | 2581 | 68 | ||
XM_008761763.1 | XP_008759985.1 | 6 - 7 | 3231 | 68 | ||
XM_008761764.1 | XP_008759986.1 | 5 - 6 | 3047 | 68 | ||
XM_008761765.1 | XP_008759987.1 | 7 - 8 | 3351 | 68 | ||
XM_008761766.1 | XP_008759988.1 | 4 - 5 | 2521 | 68 | ||
XM_008761767.1 | XP_008759989.1 | 4 - 5 | 2987 | 68 | ||
XM_008761768.1 | XP_008759990.1 | 4 - 5 | 2031 | 68 | ||
XM_008761769.1 | XP_008759991.1 | 5 - 6 | 2487 | 68 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() |
MicroArray Probes:
