Assay Details
Gene Symbol: Mmadhc | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001004280.1 | NP_001004280.1 | 5 - 6 | 585 | 86 | |
XM_006234158.2 | XP_006234220.1 | 5 - 6 | 704 | 86 | ||
GenBank mRNA | BC078953.1 | - | 5 - 6 | 585 | 86 | |
FQ212704.1 | - | 5 - 6 | 580 | 86 | ||
FQ213711.1 | - | 5 - 6 | 562 | 86 | ||
FQ216239.1 | - | 5 - 6 | 580 | 86 | ||
FQ219117.1 | - | 5 - 6 | 581 | 86 | ||
FQ228704.1 | - | 5 - 6 | 581 | 86 | ||
FQ229971.1 | - | 5 - 6 | 550 | 86 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
