Assay Details
Gene Symbol: Acvr1c | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_139090.1 | NP_620790.1 | 4 - 5 | 918 | 106 | |
GenBank mRNA | U35025.1 | - | 4 - 5 | 803 | 106 | |
U63318.1 | - | 1 - 2 | 118 | 106 | ||
U69702.1 | - | 4 - 5 | 918 | 106 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
