Assay Details
Gene Symbol: Acvr1c | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_139090.1 | NP_620790.1 | 6 - 7 | 1240 | 146 | |
GenBank mRNA | U35025.1 | - | 6 - 7 | 1125 | 146 | |
U69702.1 | - | 6 - 7 | 1240 | 146 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
