Assay Details
Gene Symbol: Zeb2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001033701.1 | NP_001028873.1 | 598 | 114 | ||
XM_006234144.2 | XP_006234206.2 | 958 | 114 | |||
XM_006234145.2 | XP_006234207.1 | 1478 | 114 | |||
XM_006234147.2 | XP_006234209.1 | 4 - 5 | 1675 | 114 | ||
XM_008761747.1 | XP_008759969.1 | 724 | 114 | |||
XM_008761748.1 | XP_008759970.1 | 957 | 114 | |||
GenBank mRNA | AM084708.1 | - | 598 | 114 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
