Assay Details
Gene Symbol: Acvr1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_024486.1 | NP_077812.1 | 7 - 8 | 1214 | 58 | |
XM_006234214.2 | XP_006234276.1 | 9 - 10 | 2819 | 58 | ||
XM_008761862.1 | XP_008760084.1 | 10 - 11 | 2876 | 58 | ||
XM_008761863.1 | XP_008760085.1 | 10 - 11 | 5120 | 58 | ||
XM_008761864.1 | XP_008760086.1 | 10 - 11 | 4866 | 58 | ||
XM_008761865.1 | XP_008760087.1 | 10 - 11 | 4909 | 58 | ||
XM_008761866.1 | XP_008760088.1 | 10 - 11 | 1586 | 58 | ||
XM_008761867.1 | XP_008760089.1 | 7 - 8 | 1130 | 58 | ||
GenBank mRNA | BC167754.1 | - | 8 - 9 | 1385 | 58 | |
L19341.1 | - | 7 - 8 | 1214 | 58 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
