Assay Details
Gene Symbol: Enoph1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001009391.1 | NP_001009391.1 | 2 - 3 | 281 | 65 | |
XM_008770023.1 | XP_008768245.1 | 2 - 3 | 331 | 65 | ||
GenBank mRNA | BC087697.1 | - | 2 - 3 | 281 | 65 |

More Information
Updated Mapping Information

Additional Information:
This assay is in the 5'UTR region of the XM_008770023.1 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
