Assay Details
Gene Symbol: Ibsp | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_012587.2 | NP_036719.2 | 5 - 6 | 325 | 89 | |
GenBank mRNA | AB001383.1 | - | 4 - 5 | 252 | 89 | |
BC127506.1 | - | 5 - 6 | 321 | 8369751 | 89 | |
DQ213013.1 | - | 4 - 5 | 252 | 89 | ||
DY314698.1 | - | 5 - 6 | 325 | 89 | ||
J04215.1 | - | 5 - 6 | 331 | 89 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
