Assay Details
Gene Symbol: Nudt9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001006991.1 | NP_001006992.1 | 3 - 4 | 537 | 71 | |
XM_006250630.2 | XP_006250692.1 | 2 - 3 | 338 | 71 | ||
XM_006250631.2 | XP_006250693.1 | 3 - 4 | 360 | 71 | ||
GenBank mRNA | BC083722.1 | - | 3 - 4 | 537 | 71 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
