Assay Details
Gene Symbol: Nudt9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001006991.1 | NP_001006992.1 | 5 - 6 | 731 | 102 | |
XM_006250630.2 | XP_006250692.1 | 4 - 5 | 532 | 102 | ||
XM_006250631.2 | XP_006250693.1 | 5 - 6 | 554 | 102 | ||
GenBank mRNA | BC083722.1 | - | 5 - 6 | 731 | 7190984 | 102 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
