Assay Details
Gene Symbol: Sparcl1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_012946.1 | NP_037078.1 | 8 - 9 | 1926 | 93 | |
GenBank mRNA | BC061755.1 | - | 8 - 9 | 1927 | 5600008 | 93 |
U27562.1 | - | 8 - 9 | 1926 | 93 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
